ContactUsLinkComponent
Human Niemann Pick C1 ELISA Kit
Human Niemann Pick C1 ELISA Kit
Catalog # ABCA_AB313912-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Human Niemann Pick C1 ELISA Kit
Catalog # ABCA_AB313912-1X96
Supplier:  Abcam
Supplier Number:  AB313912-1X96TESTS
New Product
CAS Number:  
About this item
Human Niemann Pick C1 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Niemann Pick C1 in Human in cell culture supernatant, plasma, Serum samples.