Streptavidin-HRP Solution
Streptavidin-HRP solution, 110 µl
Catalog # ABCA_AB210901-110U
New Product
Supplier: Abcam
Supplier #: AB210901-110UL
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Streptavidin-HRP Solution (ab210901) is part of the reagents, controls and accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.
Caution: RUO
Specifications
Specifications
- Description:Streptavidin-HRP solution
- Size:110 µl
- Cat. No.:ABCA_AB210901-110U
- Storage Temperature:4 °C