ContactUsLinkComponent
Canine IFN-alpha ELISA Kit
Canine IFN-alpha ELISA Kit
Catalog # ABCA_AB273185-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Canine IFN-alpha ELISA Kit
Catalog # ABCA_AB273185-1X96
Supplier:  Abcam
Supplier Number:  AB273185-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Canine IFN-alpha ELISA kit
  • Size:
    96 tests
Specifications
About this item
Canine IFN-alpha ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Canine IFN-alpha in Dog in cell culture supernatant, serum, EDTA plasma samples.