SARS-CoV-2 (COVID-19) IgA ELISA Kit
ABCA_AB277286-1X96
: Abcam
New Product
:
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:SARS-CoV-2 nucleocapsid protein
- Sample Type:Citrate plasma, Heparin Plasma, Serum
- Storage Temperature:+4 °C
- Regulatory Status:RUO