Mouse Calprotectin ELISA kit (S100A8/S100A9)
ABCA_AB267630-1X96
: Abcam
New Product
:
- Target protein:S100A8 + S100A9
Mouse Calprotectin ELISA kit (S100A8/S100A9) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Calprotectin (S100A8/S100A9) in Mouse in Plasma, Cell culture supernatant, Serum samples.