10X Wash Buffer PT
ABCA_AB206977-1000
: Abcam
New Product
:
- Description:10X Wash buffer PT
- Size:1000 ml
- Storage Temperature:4 °C
- Format:Pre-coated
10X Wash Buffer PT (ab206977) is part of the reagents, controls and accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.
: RUO