Wash Buffer Concentrate (20X)
ABCA_AB172375-30ML
: Abcam
New Product
:
- Description:Wash buffer concentrate (20X)
- Size:30 ml
- Storage Temperature:4 °C
- Format:Pre-coated
Wash Buffer Concentrate (20X) (ab172375) is part of the reagents, controls and accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.
: RUO