Complex IV Human Enzyme Activity Dipstick Assay Kit
Catalog # ABCA_AB109876-30TE
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Complex IV Human enzyme activity dipstick assay kit
- Size:30 tests
- Method:Sandwich
- Protein:Complex IV
- Species Reactivity:Other;Human
- Sample Type:Cell culture extracts, tissue
- Storage Temperature:Dual temperature (dry ice)
- Processing Time:Multiple steps
- Product Type:Pre-coated
Specifications
About this item
Complex IV Human Enzyme Activity Dipstick Assay Kit is an immunoassay kit for the detection of Complex IV Human Enzyme Activity in Cow, Human in Cell culture extracts, Tissue samples.