ContactUsLinkComponent
Phospho-ErbB3/HER3 (Y1262) and Total ErbB3/HER3 ELISA Kit
Phospho-ErbB3/HER3 (Y1262) and Total ErbB3/HER3 ELISA Kit
Catalog # ABCA_AB279791-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Phospho-ErbB3/HER3 (Y1262) and Total ErbB3/HER3 ELISA Kit
Catalog # ABCA_AB279791-1X96
Supplier:  Abcam
Supplier Number:  AB279791-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Phospho-ErbB3/HER3 (Y1262) and Total ErbB3/HER3 ELISA kit
  • Size:
    96 tests
Specifications
About this item
Phospho-ErbB3/HER3 (Y1262) and Total ErbB3/HER3 ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phospho-ErbB3/HER3 (Y1262) and Total ErbB3/HER3 in Human in Cell Lysate samples.