Human IL-12p70 ELISpot Pair
ABCA_AB281987-10X9
: Abcam
New Product
:
- Description:Human IL-12p70 ELISpot Pair
- Size:10×96 tests
- Species Reactivity:Human
- Sample Type:Suspension cells
- Form:Uncoated
- Antigen Name:IL-12 p70
- Storage Temperature:+4 °C
- Processing Time:Multiple steps
- Regulatory Status:RUO
Human IL-12p70 ELISpot Pair is a ELISpot Pair for the sensitive detection of Human IL-12p70 production in a single cell suspension.