ContactUsLinkComponent
Mouse Retinol Binding Protein ELISA Kit (RBP)
Mouse Retinol Binding Protein ELISA Kit (RBP)
Catalog # ABCA_AB190535-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Mouse Retinol Binding Protein ELISA Kit (RBP)
Catalog # ABCA_AB190535-1X96
Supplier:  Abcam
Supplier Number:  AB190535-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Mouse Retinol Binding Protein ELISA kit (RBP)
  • Size:
    96 tests
Specifications
About this item
Mouse Retinol Binding Protein ELISA Kit (RBP) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Retinol Binding Protein (RBP) in Mouse in Plasma, Serum samples.