Human Fas Ligand ELISPOT Set
Supplier: Abcam
New Product
Certificates
About this item
Human Fas Ligand ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Fas Ligand production in a single cell suspension.
Specifications
- Species Reactivity:Human
- Sample Type:Suspension cells
- Antigen Name:Fas Ligand
- Storage Temperature:4 °C
- Processing Time:Multiple steps
Specifications
Frequently Bought Together