Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
ELISAs
Phosphotyrosine c-Kit ELISA Kit
Phosphotyrosine c-Kit ELISA Kit
Phosphotyrosine c-Kit ELISA Kit
Catalog # ABCA_AB279918-1X96
Supplier:
Abcam
New Product
CAS Number:
Expand image
Phosphotyrosine c-Kit ELISA Kit
Catalog # ABCA_AB279918-1X96
Supplier:
Abcam
Supplier Number:
AB279918-1X96TESTS
New Product
CAS Number:
Specifications
Target protein:
c-Kit
Specifications
About this item
Product Details
Product Details