ContactUsLinkComponent
Phospho-Hsp27 (S82) and Total Hsp27 ELISA Kit
Phospho-Hsp27 (S82) and Total Hsp27 ELISA Kit
Catalog # ABCA_AB279823-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Phospho-Hsp27 (S82) and Total Hsp27 ELISA Kit
Catalog # ABCA_AB279823-1X96
Supplier:  Abcam
Supplier Number:  AB279823-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Phospho-Hsp27 (S82) and Total Hsp27 ELISA kit
  • Size:
    96 tests
Specifications
About this item
Phospho-Hsp27 (S82) and Total Hsp27 ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phospho-Hsp27 (S82) and Total Hsp27 in Human in Cell Lysate samples.