Canine IFN-gamma ELISA Kit (IFNG)
Catalog # ABCA_AB193684-1X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Target protein:Interferon-gamma (IFNG)
- Description:Canine IFN-gamma ELISA kit (IFNG)
- Size:96 tests
- Sample Type:Plasma, cell culture supernatant, serum
- Detection Method:Colorimetric
- Detection Range:0.1 - 25 ng/ml
- Storage Temperature:4 °C
- Sensitivity:0.1 ng/ml
- Regulatory Status:RUO
Specifications
About this item
Canine IFN-gamma ELISA kit (IFNG) is a Sandwich (quantitative) ELISA kit for the measurement of Canine IFN-gamma (IFNG) in Dog in plasma, cell culture supernatant, Serum samples.