pan-HMR GSK3 beta ELISA Kit
Catalog # ABCA_AB315067-1X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Target protein:GSK3B
- Description:pan-HMR GSK3 beta ELISA kit
- Size:96 tests
- Sample Type:Cell lysate
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Detection Range:312.5 - 20000 pg/ml
- Storage Temperature:4 °C
- Sensitivity:34.77 pg/ml
- Regulatory Status:RUO
Specifications
About this item
pan-HMR GSK3 beta ELISA kit is a single-wash 90-min Simplestep used to quantify pan-HMR GSK3 beta with a sensitivity of 34.77 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader .