ContactUsLinkComponent
Mouse G-CSF ELISA Kit (CSF3)
Mouse G-CSF ELISA Kit (CSF3)
Catalog # ABCA_AB100684-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Mouse G-CSF ELISA Kit (CSF3)
Catalog # ABCA_AB100684-1X96
Supplier:  Abcam
Supplier Number:  AB100684-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Mouse G-CSF ELISA kit (CSF3)
  • Size:
    96 tests
Specifications
About this item
Mouse G-CSF ELISA kit (CSF3) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse G-CSF (CSF3) in Mouse in plasma, cell culture supernatant, serum samples.