Phospho-TAK1 (S412) and Total TAK1 ELISA Kit
Catalog # ABCA_AB279951-1X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:TAK1
- Description:Phospho-TAK1 (S412) and Total TAK1 ELISA kit
- Size:96 tests
- Sample Type:Cell lysate
- Storage Temperature:4 °C
- Regulatory Status:RUO
Specifications
About this item
Phospho-TAK1 (S412) and Total TAK1 ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phospho-TAK1 (S412) and Total TAK1 in Human in Cell Lysate samples.