Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
ELISAs
Phospho-Smad4 (T277) and Total Smad4 ELISA Kit
Phospho-Smad4 (T277) and Total Smad4 ELISA Kit
Phospho-Smad4 (T277) and Total Smad4 ELISA Kit
Catalog # ABCA_AB279932-1X96
Supplier:
Abcam
New Product
CAS Number:
Expand image
Phospho-Smad4 (T277) and Total Smad4 ELISA Kit
Catalog # ABCA_AB279932-1X96
Supplier:
Abcam
Supplier Number:
AB279932-1X96TESTS
New Product
CAS Number:
Specifications
Target protein:
Smad4
Specifications
About this item
Product Details
Product Details