ContactUsLinkComponent
Human TPFI2 ELISA Kit
Human TPFI2 ELISA Kit
Catalog # ABCA_AB275908-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Human TPFI2 ELISA Kit
Catalog # ABCA_AB275908-1X96
Supplier:  Abcam
Supplier Number:  AB275908-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Human TPFI2 ELISA kit
  • Size:
    96 tests
Specifications
About this item
Human TPFI2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human TPFI2 with a sensitivity of 16 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.