Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sample Diluent NBS

Sample Diluent NBS

Sample diluent NBS, 20 ml
Catalog # ABCA_AB221827-20ML
New Product
Supplier:   Abcam
Supplier #:  AB221827-20ML
Sample Diluent NBS
Catalog # ABCA_AB221827-20ML
Supplier:  Abcam
New Product
CAS Number:  
Sample Diluent NBS
Sample Diluent NBS

Sample diluent NBS, 20 ml

CAS Number:  
Price
$79.10
Log in for availability
Each (20ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sample Diluent NBS (ab221827) is part of the reagents, controls and accessories range. High-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates are available.

Certifications: RUO.

Caution: RUO

Specifications

Specifications

  • Description:
    Sample diluent NBS
  • Size:
    20 ml
  • Cat. No.:
    ABCA_AB221827-20ML
  • Storage Temperature:
    4 °C

Specifications