Mouse Vitronectin ELISA Kit
Catalog # ABCA_AB272778-1X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:Vitronectin/S-Protein
- Description:Mouse Vitronectin ELISA kit
- Size:96 tests
- Detection Range:0.1 - 100 ng/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
Specifications
About this item
Mouse Vitronectin ELISA kit is a Quantitative ELISA kit for the measurement of Mouse Vitronectin in Mouse in Citrate plasma, EDTA plasma, Heparin plasma, Other biological fluids samples.