Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human KLK6 ELISA Kit

Human KLK6 ELISA Kit

Human KLK6 ELISA kit, 96 tests
Catalog # ABCA_AB193706-1X96
New Product
Supplier:   Abcam
Supplier #:  AB193706-1X96TESTS
Human KLK6 ELISA Kit
Catalog # ABCA_AB193706-1X96
Supplier:  Abcam
New Product
CAS Number:  
Human KLK6 ELISA Kit
Human KLK6 ELISA Kit

Human KLK6 ELISA kit, 96 tests

CAS Number:  
Price
$807.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human KLK6 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human KLK6 in Human in plasma, cell culture supernatant, serum samples.

Specifications

Specifications

  • Description:
    Human KLK6 ELISA kit
  • Size:
    96 tests
  • Cat. No.:
    ABCA_AB193706-1X96
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.6 ng/ml
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Human
  • Target protein:
    Kallikrein 6
  • Sample Type:
    Plasma, Cell culture supernatant, Serum
  • Detection Range:
    0.6 - 150 ng/ml
  • Detection Method:
    Colorimetric
  • Storage Temperature:
    4 °C

Specifications