Human Interferon gamma ELISPOT Kits (with un-coated plates)
Supplier: Abcam
New Product
Certificates
About this item
Human Interferon gamma ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Human Interferon gamma production in a single cell suspension.
Specifications
- Species Reactivity:Human
- Antigen Name:Interferon gamma
- Storage Temperature:4 °C
- Processing Time:Multiple steps
- Regulatory Status:RUO
Specifications
Frequently Bought Together