ContactUsLinkComponent
Human Methionine Aminopeptidase 2/p67 ELISA Kit
Human Methionine Aminopeptidase 2/p67 ELISA Kit
Catalog # ABCA_AB313992-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Human Methionine Aminopeptidase 2/p67 ELISA Kit
Catalog # ABCA_AB313992-1X96
Supplier:  Abcam
Supplier Number:  AB313992-1X96TESTS
New Product
CAS Number:  
About this item
Human Methionine Aminopeptidase 2/p67 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Methionine Aminopeptidase 2/p67 in Human in Serum, Plasma, Cell culture supernatant samples.