TMB substrate 1X
Catalog # ABCA_AB210902-120M
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:TMB substrate 1X
- Size:120 ml
- Storage Temperature:4 °C
- Plate Types:Uncoated
Specifications
About this item
TMB substrate recommended for use with the Matched Antibody Pair kits for sandwich ELISA.