ContactUsLinkComponent
Phospho-CD167a/DDR1 (Y792) and Total CD167a/DDR1 ELISA Kit
Phospho-CD167a/DDR1 (Y792) and Total CD167a/DDR1 ELISA Kit
Catalog # ABCA_AB279769-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Phospho-CD167a/DDR1 (Y792) and Total CD167a/DDR1 ELISA Kit
Catalog # ABCA_AB279769-1X96
Supplier:  Abcam
Supplier Number:  AB279769-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Phospho-CD167a/DDR1 (Y792) and Total CD167a/DDR1 ELISA kit
  • Size:
    96 tests
Specifications
About this item
Phospho-CD167a/DDR1 (Y792) and Total CD167a/DDR1 ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phospho-CD167a/DDR1 (Y792) and Total CD167a/DDR1 in Human in cell Lysate samples.