ContactUsLinkComponent
SARS-CoV-2 IgG ELISA Kit
SARS-CoV-2 IgG ELISA Kit
Catalog # ABCA_AB275300-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
SARS-CoV-2 IgG ELISA Kit
Catalog # ABCA_AB275300-1X96
Supplier:  Abcam
Supplier Number:  AB275300-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    SARS-CoV-2 IgG ELISA kit
  • Size:
    96 tests
Specifications
About this item
SARS-CoV-2 IgG ELISA kit is a Semi-quantitative ELISA kit for the measurement of SARS-CoV-2 IgG in Citrate plasma, EDTA plasma, Lithium Heparin plasma, serum samples.