Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
ELISAs
Human IFN alpha/beta R2 ELISA Kit (IFNAR2)
Human IFN alpha/beta R2 ELISA Kit (IFNAR2)
Human IFN alpha/beta R2 ELISA Kit (IFNAR2)
Catalog # ABCA_AB264610-1X96
Supplier:
Abcam
New Product
CAS Number:
Expand image
Human IFN alpha/beta R2 ELISA Kit (IFNAR2)
Catalog # ABCA_AB264610-1X96
Supplier:
Abcam
Supplier Number:
AB264610-1X96TESTS
New Product
CAS Number:
Specifications
Description:
Human IFN alpha/beta R2 ELISA kit (IFNAR2)
Size:
96 tests
Specifications
About this item
Product Details
Product Details