ContactUsLinkComponent
Mouse Insulin like Growth Factor 1 ELISA Kit (IGF1)
Mouse Insulin like Growth Factor 1 ELISA Kit (IGF1)
Catalog # ABCA_AB108874-1X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Mouse Insulin like Growth Factor 1 ELISA Kit (IGF1)
Catalog # ABCA_AB108874-1X96
Supplier:  Abcam
Supplier Number:  AB108874-1X96TESTS
New Product
CAS Number:  
Specifications
  • Description:
    Mouse Insulin like Growth Factor 1 ELISA kit (IGF1)
  • Size:
    96 tests
Specifications
About this item
Mouse Insulin like Growth Factor 1 ELISA kit (IGF1) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Insulin like Growth Factor 1 (IGF1) in Mouse in Plasma, Cell culture supernatant, Serum samples.