SARS-CoV-2 IgM ELISA Kit
Catalog # ABCA_AB275299-1X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Target protein:SARS-CoV-2 Spike Glycoprotein S1
Specifications
About this item
SARS-CoV-2 IgM ELISA Kit is a Semi-quantitative ELISA kit for the measurement of SARS-CoV-2 IgM in Citrate plasma, EDTA Plasma, Lithium Heparin Plasma, Serum samples.