Mouse Complement C5 ELISA Kit
ABCA_AB264609-1X96
: Abcam
New Product
:
- Target protein:C5
Mouse Complement C5 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Complement C5 with a sensitivity of 1.97 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.