Mouse IgG ELISA Kit
Mouse IgG ELISA kit, 96 tests
ABCA_AB157719-1X96
New Product
: Abcam
: AB157719-1X96TESTS
Mouse IgG ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IgG in Mouse in Plasma, Serum, Other biological fluids samples.
- Description:Mouse IgG ELISA kit
- Size:96 tests
- Cat. No.:ABCA_AB157719-1X96
- Regulatory Status:RUO
- Assay duration:Multiple steps
- Sensitivity:2.374 ng/ml
- Format:Pre-coated
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Target protein:IgG
- Sample Type:Plasma, Serum, Other biological fluids
- Detection Range:6.25 - 200 ng/ml
- Detection Method:Colorimetric
- Storage Temperature:+4 °C



