Rat TNF alpha ELISpot Pair
ABCA_AB281995-10X9
: Abcam
New Product
:
- Description:Rat TNF alpha ELISpot Pair
- Size:10×96 tests
- Species Reactivity:Rat
- Sample Type:Suspension cells
- Storage Temperature:4 °C
- Processing Time:Multiple steps
- Antigen Symbol:TNF alpha
- Regulatory Status:RUO
Rat TNF alpha ELISpot Pair is a ELISpot Pair for the sensitive detection of Rat TNF alpha production in a single cell suspension.