96-Well Microplate Pack
ABCA_AB210903-10PA
: Abcam
New Product
- Description:96-Well Microplate pack
- Size:10 pack
- Detection Method:Multiple steps
- Storage Temperature:+4 ℃
- Well Profile:Uncoated
96-Well Microplate Pack (ab210903) is part of the reagents, controls and accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.
: RUO