Phosphotyrosine Lck ELISA Kit
ABCA_AB279847-1X96
: Abcam
New Product
:
- Description:Phosphotyrosine Lck ELISA kit
- Size:96 tests
Phosphotyrosine Lck ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine Lck in Human in Cell Lysate samples.