SARS-CoV-2 Nucleoprotein Antibody Pair - BSA and Azide Free
Catalog # ABCA_AB309552-10X9
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:SARS-CoV-2 Nucleoprotein
- Description:SARS-CoV-2 Nucleoprotein antibody pair - BSA and azide free
- Size:10×96 tests
- Detection Range:23.44 - 3000 pg/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
Specifications
About this item
SARS-CoV-2 Nucleoprotein antibody pair - BSA and Azide free.