Pyruvate dehydrogenase (PDH) Enzyme Activity Dipstick Assay Kit
Catalog # ABCA_AB109882-30TE
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Pyruvate dehydrogenase (PDH) enzyme activity dipstick assay kit
- Size:30 tests
- Method:Sandwich
- Protein:Pyruvate dehydrogenase
- Species Reactivity:Other, Human, Mouse, Pig, Rat
- Sample Type:Cell culture extracts, tissue
- Storage Temperature:Dual temperature (Dry ice)
- Processing Time:Multiple steps
- Product Type:Pre-coated
- Format:Pre-coated
Specifications
About this item
Pyruvate dehydrogenase (PDH) Enzyme Activity Dipstick Assay kit is an immunoassay kit for the detection of Pyruvate dehydrogenase (PDH) Enzyme Activity in Mouse, Rat, Cow, Human, Pig in cell culture extracts, Tissue samples.
Caution:
RUO