HRP Stability Conjugate Reagent
: Abcam
New Product
Abcam's HRP Stability Conjugate Reagent (ab171537) was developed specifically for Horseradish Peroxidase (HRP) conjugates in solution.
: RUO
:
RUO
- Form:Liquid
- Storage Temperature:Store at room temperature
- Application:ELISA buffer
Frequently Bought Together