Rat Interferon gamma ELISpot Pair
ABCA_AB281994-10X9
: Abcam
New Product
:
- Description:Rat Interferon gamma ELISpot Pair
- Size:10×96 tests
- Species Reactivity:Rat
- Sample Type:Suspension cells
- Form:Uncoated
- Storage Temperature:+4 °C
- Processing Time:Multiple steps
- Antigen Symbol:Interferon gamma
- Regulatory Status:RUO
Rat Interferon gamma ELISpot Pair is a ELISpot Pair for the sensitive detection of Rat Interferon gamma production in a single cell suspension.