Phospho-PKMYT1 (T495) and Total PKMYT1 ELISA Kit
ABCA_AB279891-1X96
: Abcam
New Product
:
- Description:Phospho-PKMYT1 (T495) and Total PKMYT1 ELISA kit
- Size:96 tests
Phospho-PKMYT1 (T495) and Total PKMYT1 ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phospho-PKMYT1 (T495) and Total PKMYT1 in Human in Cell Lysate samples.