Mouse Interferon gamma ELISPOT Kits
Supplier: Abcam
New Product
Certificates
About this item
Mouse Interferon gamma ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Mouse Interferon gamma production in a single cell suspension.
Specifications
- Species Reactivity:Mouse
- Antigen Name:Interferon gamma
- Storage Temperature:4 °C
- Processing Time:Multiple steps
- Regulatory Status:RUO
Specifications
Frequently Bought Together