SARS-CoV2 Nucleoprotein ELISA Kit
Catalog # ABCA_AB315296-1X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Target protein:SARS Nucleocapsid Protein
- Sample Type:Serum, EDTA Plasma, Saliva, Cell culture media
- Detection Method:Colorimetric
- Time to Results:1 hr 30 min
- Detection Range:46.875 - 3000 pg/ml
- Storage Temperature:+4 °C
- Sensitivity:17.045 pg/ml
- Regulatory Status:RUO
Specifications
About this item
SARS-CoV2 Nucleoprotein ELISA Kit is a single-wash 90-min Simplestep used to quantify SARS-CoV2 Nucleoprotein with a sensitivity of 17.045 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader .