ContactUsLinkComponent
Antibody enhancer
Antibody enhancer
Catalog # ALA494276100ML
Supplier:  ALADDIN SCIENTIFIC
New Product
CAS Number:  
Antibody enhancer
Catalog # ALA494276100ML
Supplier:  ALADDIN SCIENTIFIC
Supplier Number:  A494276-100ML
New Product
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pack type:
    Glass bottle
  • Description:
    Antibody Enhancer
  • Size:
    100 ml
Specifications
About this item
Antibody enhancer, ready-to-use (RTU)
  • The product secures molecular conformation and prevents loss of activity of the diluent protein, over time. Antibody Enhancer enables users to create prediluted ready-to-use conjugated antibodies (free poly- and monoclonal antibodies, HRP-conjugated antibodies, AP-conjugated antibodies and fluorescence-conjugated antibodies): Reduces background and provides highest signal-to-noise ratio due to the antibody’s enhanced binding capacity.  Amplifies 40-80% the specific antibody’s activity without increasing the non-specific binding.  Allows for the diluted conjugate to be stored at least 4 years at 2…8 °C.  Minimizes assay errors related to daily dilution variation.  Limited or expensive conjugates can be further diluted without losing sensitivity.  Designed for use in colorimetric as well as in chemiluminescence based assays such as ELISA, Western Blotting and IHC. The buffer is ready-to-use and BSA free. It contains no azide, mercury or other toxic preservative formulations.
Antibody Enhancer is a signal enhancing buffer that can be used for the dilution, stabilization and binding capacity enhancement of concentrated antibodies and conjugated antibodies. The product secures molecular conformation and prevents loss of activity, of the diluent protein, over time. Antibody Enhancer enables users to create prediluted ready-to-use conjugated antibodies (free poly- and monoclonal antibodies, HRP-conjugated antibodies, AP-conjugated antibodies and fluorescence-conjugated antibodies): Reduces background and provides highest signal-to-noise ratio due to the antibody’s enhanced binding capacity. Amplifies 40-80% the specific antibody’s activity without increasing the non-specific binding. Allows for the diluted conjugate to be stored at least 4 years at 2…8 °C. Minimizes assay errors related to daily dilution variation. Limited or expensive conjugates can be further diluted without losing sensitivity. Designed for use in colorimetric as well as in chemiluminescence based assays such as ELISA, Western Blotting and IHC.
Working Procedure 1.Make a series of dilutions of your antibody/antibody conjugate in the Antibody Enhancer in order to determine the optimal dilution factor (the enhancement is significant in dilutions higher than 1:2000, compared to HRP-StabilPLUS (cat. no. H494387)). 2.Run the assay as usual or store the diluted, conjugated protein preferably at 2…8 °C. Tips and Tricks  Antibody Enhancer cannot be used for the dilution of antibodies directed against rabbit immunoglobulins i.e. Swine anti-Rabbit Ig and Goat anti-Rabbit Ig etc. UNI-StabilPLUS (cat. no. U494463) is recommended for this. Handling and Storage  Store solution at 2…8 °C.
Packaging: Refrigerator