Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Plastic bottle
- CAS No.:56-81-5
- Size:1 gal
- Storage Conditions:Store at RT
- MDL No.:MFCD00004722
- Protein/Peptide Name:Glycerol
- Purity:Purity by GC ≥ 99.5%
- Molecular Weight:92,09 g/mol
- Grade:ACS
- Molecular Formula:C₃H₈O₃
Glycerol, also known as glycerin or glycerine, is a versatile and widely used compound in various industries due to its unique properties. This colorless, odorless liquid is hygroscopic, meaning it can attract and retain moisture, making it an excellent humectant in cosmetic formulations, food products, and pharmaceuticals. Glycerol is commonly utilized in the production of personal care items such as lotions and creams, where it helps to maintain skin hydration and improve texture. In the food industry, it serves as a sweetener and preservative, enhancing the shelf life and palatability of products. Moreover, glycerol plays a crucial role in the pharmaceutical sector, where it is used as a solvent and a vehicle for drug delivery. Its non-toxic nature and compatibility with various substances make it ideal for formulating syrups and elixirs. Additionally, glycerol is employed in the production of biodiesel, where it acts as a byproduct of the transesterification process, showcasing its potential in sustainable energy solutions. With its multifunctional applications and safety profile, glycerol remains an essential ingredient across multiple sectors.
Density: 1,26 g/cm³
Boiling Point: 290 °C (Approx.)