Order Entry
Puerto Rico
ContactUsLinkComponent
N-Formyl-Met-Leu-Phe
N-Formyl-Met-Leu-Phe
Supplier:  ALADDIN SCIENTIFIC
New Product
Certificates
N-Formyl-Met-Leu-Phe
Supplier:  ALADDIN SCIENTIFIC
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    59880-97-6
  • Storage Conditions:
    Freezer
  • MDL No.:
    MFCD00036780
  • Protein Synonyms:
    Chemotactic peptide fmlF fmlP (misnomer but widely used) N-Formyl-L-methionyl-L-leucyl-L-phenylalanine
  • Protein/Peptide Name:
    N-Formyl-Met-Leu-Phe
  • Purity:
    ≥97%
  • Molecular Weight:
    437.55 g/mol
  • Grade:
    Moligand™
  • Molecular Formula:
    C₂₁H₃₁N₃O₅S

Specifications