Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Plastic bottle
- CAS No.:9048-46-8
- Source:Blood
- Species:Bovine
- Size:25 g
- Storage Conditions:Store at 0 - 8 °C
- MDL No.:MFCD00145743
- Protein/Peptide Name:Albumin fraction V
- Purity:Purity ≥ 97% (Electrophoresis)
Albumin fraction V is a highly purified protein derived from serum albumin, recognized for its exceptional binding properties and stability. This compound is widely utilized in various biotechnological and pharmaceutical applications, particularly in drug formulation and delivery systems. Its unique ability to bind to a wide range of molecules, including fatty acids and hormones, makes it an invaluable tool in research and clinical settings. Researchers often leverage Albumin fraction V for its role in stabilizing proteins and enhancing the solubility of hydrophobic compounds, thereby improving the efficacy of therapeutic agents. In addition to its use in drug delivery, Albumin fraction V serves as a standard in protein assays and is employed in the development of diagnostic tests. Its biocompatibility and low immunogenicity further enhance its appeal in medical applications, including tissue engineering and regenerative medicine. With its versatile applications and significant advantages over other protein carriers, Albumin fraction V stands out as a critical component in advancing both research and therapeutic innovations.