Ham's F10
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
New Product
Certificates
About this item
Ham's F10 was originally developed for use in cloning Chinese hamster ovary and lung cells. This medium is useful with serum supplements for a variety of cell culture applications, including the growth of myeloma and hybridoma cells.
Animal component-free
Chemically defined
GMP manufacture
With: 1200 mg/L Sodium bicarbonate
Without: L-Glutamine - add 5 ml/L (200 mM)
Specifications
- Calcium:Yes
- Magnesium:Yes
- HEPES:No
- Sodium bicarbonate:Yes
- Sodium pyruvate:Yes
- Phenol red:Yes
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:High
- Media Base:Ham's F10 Nutrient Mixture