Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Trypsin EDTA 10X Solution

Trypsin EDTA 10X Solution

Liquid, 100 ml
Catalog # 77975-387
New Product
Supplier #:  9342-100ML
Trypsin EDTA 10X Solution
Catalog # 77975-387
New Product
CAS Number:  
Trypsin EDTA 10X Solution
Trypsin EDTA 10X Solution

Liquid, 100 ml

CAS Number:  
Price
$48.75
Log in for availability
Each(100ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Does not contain phenol red.

  • Animal-component free

    Chemically defined

    GMP manufacture

Trypsin EDTA 10X Solution is a combined reagent used to release adherent cells from the culture vessel surface into suspension. This formulation contains 5 g/L trypsin (1:250) and 2 g/L EDTA tetrasodium salt in Hanks' Balanced Salt Solution.

Packaging: Store at ≤−10 °C. Shelf life: 24 months from date of manufacture.

Specifications

Specifications

  • Media Format:
    Liquid
  • Size:
    100 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.