Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Media Format:Liquid
- Size:500 ml
- Calcium:Yes
- Magnesium:Yes
- HEPES:No
- Sodium bicarbonate:Yes
- Sodium pyruvate:No
- Phenol red:Yes
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:High
- Media Base:McCoy's 5A Medium
McCoy’s 5A medium is a modification of Basal Medium Eagle (BME) with changes in amino acid compositions to support optimal hepatoma cultures. Containing additional vitamins, high phosphate, and Bacto peptone, McCoy’s 5A medium has been demonstrated to support primary cell cultures from many normal adult tissues, including bone marrow, testes, kidney, skin, and gingiva.
Animal component-free
Chemically defined
GMP manufacture
McCoy's Medium 5A is a general medium for primary and established cell lines. It contains increased glucose, additional vitamins, high phosphate, and peptone. With: 3000 mg/L Glucose 2200 mg/L Sodium bicarbonate Without: L-Glutamine, add 7.51 ml/L (200 mM)